missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxyindole 98.0+%, TCI America™

Catalog No. M17515G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17515G 5 g
M17511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17515G Supplier TCI America Supplier No. M17515G

Chemical Identifiers

CAS 3189-22-8
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00047203
InChI Key FSOPPXYMWZOKRM-UHFFFAOYSA-N
Synonym 7-methoxyindole, 7-methoxy indole, 1h-indole, 7-methoxy, indole, 7-methoxy, methoxyindole 7-, unii-9r05qk9rp4, 7-methoxy-indole, pubchem7434, acmc-209hpi
PubChem CID 76660
IUPAC Name 7-methoxy-1H-indole
SMILES COC1=CC=CC2=C1NC=C2

Specifications

Boiling Point 166°C
Quantity 5 g
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 7-Methoxyindole