missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Methoxyflavone 98.0+%, TCI America™
$95.06 - $294.52
Chemical Identifiers
| CAS | 22395-22-8 |
|---|---|
| Molecular Formula | C16H12O3 |
| Molecular Weight (g/mol) | 252.269 |
| MDL Number | MFCD00017462 |
| InChI Key | QKNDCRMJDZLFEG-UHFFFAOYSA-N |
| Synonym | 7-methoxyflavone, 7-methoxy-2-phenyl-4h-chromen-4-one, flavone, 7-methoxy, unii-xk561ap5ur, 7-methoxy-2-phenyl-chromen-4-one, xk561ap5ur, 7-methoxy-2-phenylchromone, 7-methoxy flavone, 7-methoxy-2-phenyl-4h-benzopyran-4-one |
| PubChem CID | 466268 |
| IUPAC Name | 7-methoxy-2-phenylchromen-4-one |
| SMILES | COC1=CC2=C(C=C1)C(=O)C=C(O2)C3=CC=CC=C3 |
Chemical Identifiers
| 22395-22-8 | |
| 252.269 | |
| QKNDCRMJDZLFEG-UHFFFAOYSA-N | |
| 466268 | |
| COC1=CC2=C(C=C1)C(=O)C=C(O2)C3=CC=CC=C3 |
| C16H12O3 | |
| MFCD00017462 | |
| 7-methoxyflavone, 7-methoxy-2-phenyl-4h-chromen-4-one, flavone, 7-methoxy, unii-xk561ap5ur, 7-methoxy-2-phenyl-chromen-4-one, xk561ap5ur, 7-methoxy-2-phenylchromone, 7-methoxy flavone, 7-methoxy-2-phenyl-4h-benzopyran-4-one | |
| 7-methoxy-2-phenylchromen-4-one |
Specifications
| 22395-22-8 | |
| White-Yellow | |
| MFCD00017462 | |
| 7-methoxyflavone, 7-methoxy-2-phenyl-4h-chromen-4-one, flavone, 7-methoxy, unii-xk561ap5ur, 7-methoxy-2-phenyl-chromen-4-one, xk561ap5ur, 7-methoxy-2-phenylchromone, 7-methoxy flavone, 7-methoxy-2-phenyl-4h-benzopyran-4-one | |
| COC1=CC2=C(C=C1)C(=O)C=C(O2)C3=CC=CC=C3 | |
| 252.269 | |
| 252.27 | |
| Crystalline Powder |
| 111°C | |
| C16H12O3 | |
| 1 g | |
| QKNDCRMJDZLFEG-UHFFFAOYSA-N | |
| 7-methoxy-2-phenylchromen-4-one | |
| 466268 | |
| ≥98.0% (GC) | |
| 7-Methoxyflavone |
Safety and Handling
TSCA : No