missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxycoumarin 98.0+%, TCI America™

Catalog No. p-7132824
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17235G 5 g
M172325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17235G Supplier TCI America Supplier No. M17235G
Only null left

Chemical Identifiers

CAS 531-59-9
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00006876
InChI Key LIIALPBMIOVAHH-UHFFFAOYSA-N
Synonym 7-methoxycoumarin, herniarin, ayapanin, methylumbelliferone, 7-methoxy-2h-chromen-2-one, herniarine, 7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 7-methoxy, coumarin, 7-methoxy, methoxycourmarin, 7
PubChem CID 10748
ChEBI CHEBI:5679
IUPAC Name 7-methoxychromen-2-one
SMILES COC1=CC2=C(C=C1)C=CC(=O)O2

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 5 g
Formula Weight 176.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methoxycoumarin
RTECSNumber DJ3100380
TSCA Yes