Learn More
7-Methoxycoumarin, 98+%
CAS: 531-59-9 | C10H8O3 | 176.171 g/mol
Supplier: Thermo Scientific Chemicals A1544114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications7-Methoxycoumarin is a compound biosynthesized in lavender that exhibits antimicrobial activity.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
| 7-Methoxycoumarin | |
| 117°C to 120°C | |
| MFCD00006876 | |
| 141728 | |
| Insoluble in water. | |
| COC1=CC2=C(C=C1)C=CC(=O)O2 | |
| 176.171 | |
| CHEBI:5679 | |
| ≥98% |
| 531-59-9 | |
| C10H8O3 | |
| 25 g | |
| 7-methoxycoumarin, herniarin, ayapanin, methylumbelliferone, 7-methoxy-2h-chromen-2-one, herniarine, 7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 7-methoxy, coumarin, 7-methoxy, methoxycourmarin, 7 | |
| LIIALPBMIOVAHH-UHFFFAOYSA-N | |
| 7-methoxychromen-2-one | |
| 10748 | |
| 176.17 |
Chemical Identifiers
| 531-59-9 | |
| 176.171 | |
| LIIALPBMIOVAHH-UHFFFAOYSA-N | |
| 10748 | |
| 7-methoxychromen-2-one |
| C10H8O3 | |
| MFCD00006876 | |
| 7-methoxycoumarin, herniarin, ayapanin, methylumbelliferone, 7-methoxy-2h-chromen-2-one, herniarine, 7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 7-methoxy, coumarin, 7-methoxy, methoxycourmarin, 7 | |
| CHEBI:5679 | |
| COC1=CC2=C(C=C1)C=CC(=O)O2 |
Safety and Handling
EINECSNumber : 208-513-3
RTECSNumber : DJ3100380
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only