missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxycoumarin, 98+%

Catalog No. AAA1544122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1544122 100 g
AAA1544106 5 g
AAA1544114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1544122 Supplier Thermo Scientific Chemicals Supplier No. A1544122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 531-59-9 | C10H8O3 | 176.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
7-Methoxycoumarin is a compound biosynthesized in lavender that exhibits antimicrobial activity.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 531-59-9
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00006876
InChI Key LIIALPBMIOVAHH-UHFFFAOYSA-N
Synonym 7-methoxycoumarin, herniarin, ayapanin, methylumbelliferone, 7-methoxy-2h-chromen-2-one, herniarine, 7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 7-methoxy, coumarin, 7-methoxy, methoxycourmarin, 7
PubChem CID 10748
ChEBI CHEBI:5679
IUPAC Name 7-methoxychromen-2-one
SMILES COC1=CC2=C(C=C1)C=CC(=O)O2

Specifications

Melting Point 117°C to 120°C
Quantity 100 g
Beilstein 141728
Solubility Information Insoluble in water.
Formula Weight 176.17
Percent Purity ≥98%
Chemical Name or Material 7-Methoxycoumarin
EINECSNumber 208-513-3
RTECSNumber DJ3100380
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only