missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxyindole, 95%

Catalog No. AC462292500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462292500 250 mg
AC462290010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462292500 Supplier Thermo Scientific Chemicals Supplier No. 462292500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2380-84-9 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2380-84-9
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
InChI Key ORVPXPKEZLTMNW-UHFFFAOYSA-N
Synonym 7-hydroxyindole, 7-indolol, 7-hydroxy-1h-indole, indol-7-ol, 7-hydroxy-indole, pubchem7345, 7-hydroxylindole
PubChem CID 2737651
IUPAC Name 1H-indol-7-ol
SMILES C1=CC2=C(C(=C1)O)NC=C2

Specifications

Melting Point 85.0°C to 87.0°C
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 133.15
Percent Purity 95%
Chemical Name or Material 7-Hydroxyindole
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only