missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Hydroxyflavone 97.0+%, TCI America™
$106.25 - $260.97
Chemical Identifiers
| CAS | 6665-86-7 |
|---|---|
| Molecular Formula | C15H10O3 |
| Molecular Weight (g/mol) | 238.242 |
| MDL Number | MFCD00006835 |
| InChI Key | MQGPSCMMNJKMHQ-UHFFFAOYSA-N |
| Synonym | 7-hydroxyflavone, flavone, 7-hydroxy, 7-hydroxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl, 7-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 7-hydroxy, unii-ze72458e4l, 7-hydroxy-2-phenyl-chromen-4-one, 7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 7-hydroxyflavone, 11 |
| PubChem CID | 5281894 |
| ChEBI | CHEBI:2268 |
| IUPAC Name | 7-hydroxy-2-phenylchromen-4-one |
| SMILES | C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O |
Chemical Identifiers
| 6665-86-7 | |
| 238.242 | |
| MQGPSCMMNJKMHQ-UHFFFAOYSA-N | |
| 5281894 | |
| 7-hydroxy-2-phenylchromen-4-one |
| C15H10O3 | |
| MFCD00006835 | |
| 7-hydroxyflavone, flavone, 7-hydroxy, 7-hydroxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl, 7-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 7-hydroxy, unii-ze72458e4l, 7-hydroxy-2-phenyl-chromen-4-one, 7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 7-hydroxyflavone, 11 | |
| CHEBI:2268 | |
| C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O |
Specifications
| 6665-86-7 | |
| Yellow | |
| MFCD00006835 | |
| 7-hydroxyflavone, flavone, 7-hydroxy, 7-hydroxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl, 7-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 7-hydroxy, unii-ze72458e4l, 7-hydroxy-2-phenyl-chromen-4-one, 7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 7-hydroxyflavone, 11 | |
| C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O | |
| 238.242 | |
| CHEBI:2268 | |
| ≥97.0% (T) | |
| 7-Hydroxyflavone |
| 246°C | |
| C15H10O3 | |
| 1 g | |
| MQGPSCMMNJKMHQ-UHFFFAOYSA-N | |
| 7-hydroxy-2-phenylchromen-4-one | |
| 5281894 | |
| 238.24 | |
| Crystalline Powder |
Safety and Handling
TSCA : No