missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxyflavone 97.0+%, TCI America™

Catalog No. H08525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08525G 5 g
H08521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08525G Supplier TCI America Supplier No. H08525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6665-86-7
Molecular Formula C15H10O3
Molecular Weight (g/mol) 238.242
MDL Number MFCD00006835
InChI Key MQGPSCMMNJKMHQ-UHFFFAOYSA-N
Synonym 7-hydroxyflavone, flavone, 7-hydroxy, 7-hydroxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl, 7-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 7-hydroxy, unii-ze72458e4l, 7-hydroxy-2-phenyl-chromen-4-one, 7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 7-hydroxyflavone, 11
PubChem CID 5281894
ChEBI CHEBI:2268
IUPAC Name 7-hydroxy-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O

Specifications

Melting Point 246°C
Color Yellow
Quantity 5 g
Formula Weight 238.24
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxyflavone