missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxyflavone, 98%

Catalog No. p-7024215
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1853803 1 g
AAA1853806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1853803 Supplier Thermo Scientific Chemicals Supplier No. A1853803
Only null left

CAS: 6665-86-7 | C15H10O3 | 238.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6665-86-7
Molecular Formula C15H10O3
Molecular Weight (g/mol) 238.242
MDL Number MFCD00006835
InChI Key MQGPSCMMNJKMHQ-UHFFFAOYSA-N
Synonym 7-hydroxyflavone, flavone, 7-hydroxy, 7-hydroxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl, 7-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 7-hydroxy, unii-ze72458e4l, 7-hydroxy-2-phenyl-chromen-4-one, 7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 7-hydroxyflavone, 11
PubChem CID 5281894
ChEBI CHEBI:2268
IUPAC Name 7-hydroxy-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O

Specifications

Melting Point 245°C to 247°C
Quantity 1 g
Beilstein 194089
Formula Weight 238.25
Percent Purity 98%
Chemical Name or Material 7-Hydroxyflavone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 229-705-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only