missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine, TCI America™

Catalog No. H027325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H027325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H027325G Supplier TCI America Supplier No. H027325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2503-56-2
Molecular Formula C6H6N4O
Molecular Weight (g/mol) 150.141
MDL Number MFCD00005555
InChI Key INVVMIXYILXINW-UHFFFAOYSA-N
Synonym 5-methyl-1,2,4 triazolo 1,5-a pyrimidin-7-ol, 7-hydroxy-5-methyl-1,3,4-triazaindolizine, methyl hydroxytriazaindolizine, 1,2,4 triazolo 1,5-a pyrimidin-7-ol, 5-methyl, 5-methyl-7-hydroxy-1,3,4-triazaindolizine, 5-methyl-1h-1,2,4 triazolo 1,5-a pyrimidin-7-one, 5-methyl-s-triazolo 1,5-a pyrimidin-7-ol, s-triazolo 1,5-a pyrimidin-7-ol, 5-methyl, 5-methyl-1,2,4-triazolo 1,5-a pyrimidin-7-ol, 5-methyl 1,2,4 triazolo 1,5-a pyrimidin-7-ol
PubChem CID 75629
IUPAC Name 5-methyl-1H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one
SMILES CC1=CC(=O)N2C(=N1)N=CN2

Specifications

Melting Point 226°C
Color White
Quantity 25 g
Formula Weight 150.14
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxy-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine
RTECSNumber XZ6133300
TSCA Yes