missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine, 98%

Catalog No. p-7024167
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1843814 25 g
AAA1843822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1843814 Supplier Thermo Scientific Chemicals Supplier No. A1843814
Only null left

CAS: 2503-56-2 | C6H6N4O | 150.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 2503-56-2
Molecular Formula C6H6N4O
MDL Number MFCD00005555
InChI Key INVVMIXYILXINW-UHFFFAOYSA-N
Synonym 5-methyl-1,2,4 triazolo 1,5-a pyrimidin-7-ol, 7-hydroxy-5-methyl-1,3,4-triazaindolizine, methyl hydroxytriazaindolizine, 1,2,4 triazolo 1,5-a pyrimidin-7-ol, 5-methyl, 5-methyl-7-hydroxy-1,3,4-triazaindolizine, 5-methyl-1h-1,2,4 triazolo 1,5-a pyrimidin-7-one, 5-methyl-s-triazolo 1,5-a pyrimidin-7-ol, s-triazolo 1,5-a pyrimidin-7-ol, 5-methyl, 5-methyl-1,2,4-triazolo 1,5-a pyrimidin-7-ol, 5-methyl 1,2,4 triazolo 1,5-a pyrimidin-7-ol
PubChem CID 75629

Specifications

Melting Point 281°C to 285°C
Quantity 25 g
Beilstein 609440
SMILES CC1=CC(=O)N2C(=N1)N=CN2
IUPAC Name 5-methyl-1H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one
Molecular Weight (g/mol) 150.141
Formula Weight 150.14
Percent Purity 98%
Chemical Name or Material 7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-706-7
RTECSNumber X26133300
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only