missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-1-indanone, 97%

Catalog No. p-7057309
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66850MD 250 mg
AAH6685003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66850MD Supplier Thermo Scientific Chemicals Supplier No. H66850MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6968-35-0 | C9H8O2 | 148.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6968-35-0
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD01548387
InChI Key HFMZPBSZKCDKOR-UHFFFAOYSA-N
Synonym 7-hydroxy-1-indanone, 7-hydroxy-2,3-dihydro-1h-inden-1-one, 7-hydroxyindan-1-one, 1h-inden-1-one,2,3-dihydro-7-hydroxy, 4h-inden-4-one, 1,2-dihydro-3-hydroxy, 7-oxidanyl-2,3-dihydroinden-1-one, 7-hydroxyindanone, acmc-20mvbk, pubchem23254, acmc-209wv8
PubChem CID 248078
IUPAC Name 7-hydroxy-2,3-dihydro-1H-inden-1-one
SMILES OC1=CC=CC2=C1C(=O)CC2

Specifications

Melting Point 109°C to 113°C
Quantity 250 mg
Beilstein 11085807
Formula Weight 148.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 7-Hydroxy-1-indanone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only