missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-1,2,3,4-tetrahydroquinoline, 96%

Catalog No. AC458710010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458710010 1 g
AC458710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458710010 Supplier Thermo Scientific Chemicals Supplier No. 458710010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58196-33-1 | C9H11NO | 149.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58196-33-1
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
InChI Key HJJRGZMJZDSMDB-UHFFFAOYSA-N
Synonym 7-hydroxy-1,2,3,4-tetrahydroquinoline, 7-quinolinol, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro-7-quinolinol, 1,2,3,4-tetrahydro-quinolin-7-ol, 1,2,3,4-tetrahydrochinolin-7-ol, pubchem12787, 7-hydroxy-3,4-dihydro-2h-quinoline, 1,2,3,4-tetrahydro-7-hydroxyquinoline, 7-hydroxy-1,2,3,4-tetrahydro-quinolin, 7-hydroxy-1,2,3,4-tetrahydro quinolinoe
PubChem CID 93980
IUPAC Name 1,2,3,4-tetrahydroquinolin-7-ol
SMILES C1CC2=C(C=C(C=C2)O)NC1

Specifications

Melting Point 91.0°C to 93.5°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 149.19
Percent Purity 96%
Chemical Name or Material 7-Hydroxy-1,2,3,4-tetrahydroquinoline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only