missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoroisatin, 97%

Catalog No. AAH2682703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2682703 1 g
AAH2682706 5 g
AAH2682714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2682703 Supplier Thermo Scientific Chemicals Supplier No. H2682703
Only null left

CAS: 317-20-4 | C8H4FNO2 | 165.123 g/mol

7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.

Solubility
Sparingly Soluble in water (0.022 g/L)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 317-20-4
Molecular Formula C8H4FNO2
Molecular Weight (g/mol) 165.123
MDL Number MFCD01569508
InChI Key HGBGVEOXPHGSOS-UHFFFAOYSA-N
Synonym 7-fluoroisatin, 7-fluoroindoline-2,3-dione, 7-fluoro-2,3-indolinedione, 1h-indole-2,3-dione, 7-fluoro, 7-fluoroindole-1h-2,3-dione, 7-fluoroindole-2,3-dione, 7-fluoro-2,3-dihydro-1h-indole-2,3-dione, 7-fluoroisatine, 7-fluoro-1h-benzo d azoline-2,3-dione, 7-fluor-1h-indol-2,3-dion
PubChem CID 586418
IUPAC Name 7-fluoro-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)F)NC(=O)C2=O

Specifications

Melting Point 193°C to 195°C
Quantity 1 g
Solubility Information Sparingly Soluble in water (0.022g/L)
Formula Weight 165.12
Percent Purity 97%
Chemical Name or Material 7-Fluoroisatin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only