missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoroindole 98.0+%, TCI America™

Catalog No. F05551G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05551G Supplier TCI America Supplier No. F05551G

Chemical Identifiers

CAS 387-44-0
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD01074502
InChI Key XONKJZDHGCMRRF-UHFFFAOYSA-N
Synonym 7-fluoroindole, 1h-indole, 7-fluoro, pubchem7294, 7-fluoranyl-1h-indole, acmc-209j1q, ksc222e4r, 7-fluoro-1h-indole, buttpark 24\07-35
PubChem CID 2774504
IUPAC Name 7-fluoro-1H-indole
SMILES FC1=C2NC=CC2=CC=C1

Specifications

Melting Point 67°C
Quantity 1 g
Formula Weight 135.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Fluoroindole