missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoroindole, 97%

Catalog No. AAL17621MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17621MD 250 mg
AAL1762103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL17621MD Supplier Thermo Scientific Chemicals Supplier No. L17621MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 387-44-0 | C8H6FN | 135.14 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in methanol. (almost transparency)

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 387-44-0
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD01074502
InChI Key XONKJZDHGCMRRF-UHFFFAOYSA-N
Synonym 7-fluoroindole, 1h-indole, 7-fluoro, pubchem7294, 7-fluoranyl-1h-indole, acmc-209j1q, ksc222e4r, 7-fluoro-1h-indole, buttpark 24\07-35
PubChem CID 2774504
IUPAC Name 7-fluoro-1H-indole
SMILES FC1=C2NC=CC2=CC=C1

Specifications

Melting Point 61°C to 63°C
Quantity 250 mg
Beilstein 114740
Solubility Information Soluble in methanol. (almost transparency)
Formula Weight 135.14
Percent Purity 97%
Chemical Name or Material 7-Fluoroindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only