missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, 99%, Thermo Scientific Chemicals
$933.85 - $933.85
Chemical Identifiers
| CAS | 84806-27-9 |
|---|---|
| Molecular Formula | C6H6FN3O4S |
| Molecular Weight (g/mol) | 235.189 |
| MDL Number | MFCD02682109 |
| InChI Key | JXLHNMVSKXFWAO-UHFFFAOYSA-N |
| Synonym | sbd-f, 7-fluorobenzofurazan-4-sulfonic acid ammonium salt, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate, azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane, 2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1 |
| PubChem CID | 2737780 |
| IUPAC Name | azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate |
| SMILES | C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB21767MF
|
Thermo Scientific Chemicals
B21767MF |
50 mg |
Each for $933.85
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 84806-27-9 | |
| 235.189 | |
| JXLHNMVSKXFWAO-UHFFFAOYSA-N | |
| 2737780 | |
| C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+] |
| C6H6FN3O4S | |
| MFCD02682109 | |
| sbd-f, 7-fluorobenzofurazan-4-sulfonic acid ammonium salt, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate, azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane, 2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1 | |
| azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate |
Specifications
| 84806-27-9 | |
| C6H6FN3O4S | |
| 50 mg | |
| sbd-f, 7-fluorobenzofurazan-4-sulfonic acid ammonium salt, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate, azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane, 2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1 | |
| C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+] | |
| 235.189 | |
| 235.19 | |
| 7-Fluorobenzofurazan-4-sulfonic acid ammonium salt |
| ∼285°C to 295°C (decomposition) | |
| MFCD02682109 | |
| Light sensitive | |
| JXLHNMVSKXFWAO-UHFFFAOYSA-N | |
| azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate | |
| 2737780 | |
| 99% |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only