missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoro-2-methylquinoline 98.0+%, TCI America™

Catalog No. F05195G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05195G 5 g
F05191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05195G Supplier TCI America Supplier No. F05195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1128-74-1
Molecular Formula C10H8FN
Molecular Weight (g/mol) 161.179
InChI Key AYSKNDXTWPNRKG-UHFFFAOYSA-N
Synonym 7-Fluoroquinaldine
PubChem CID 2779075
IUPAC Name 7-fluoro-2-methylquinoline
SMILES CC1=NC2=C(C=C1)C=CC(=C2)F

Specifications

Melting Point 47°C
Boiling Point 120°C
Quantity 5 g
Formula Weight 161.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Fluoro-2-methylquinoline