missing translation for 'onlineSavingsMsg'
Learn More

7-Ethylindole, 98+%

Catalog No. AAL1487509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1487509 10 g
AAL1487518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1487509 Supplier Thermo Scientific Chemicals Supplier No. L1487509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 22867-74-9 | C10H11N | 145.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22867-74-9
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.21
MDL Number MFCD00143514
InChI Key PIIZLMYXLGYWTN-UHFFFAOYSA-N
Synonym 7-ethylindole, 1h-indole, 7-ethyl, 7-ethyl indole, pubchem7338, acmc-1cfby, 7-ethylindole gc
PubChem CID 2724711
IUPAC Name 7-ethyl-1H-indole
SMILES CCC1=C2NC=CC2=CC=C1

Specifications

Density 1.058
Boiling Point 230°C
Refractive Index 1.603
Quantity 10 g
Beilstein 1238356
Formula Weight 145.21
Percent Purity ≥98%
Chemical Name or Material 7-Ethylindole
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only