missing translation for 'onlineSavingsMsg'
Learn More

7-Ethylcamptothecin 96.0+%, TCI America™

Catalog No. E0781100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0781100MG 100 mg
E07811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0781100MG Supplier TCI America Supplier No. E0781100MG

Chemical Identifiers

CAS 78287-27-1
Molecular Formula C22H20N2O4
Molecular Weight (g/mol) 376.412
MDL Number MFCD06657922
InChI Key MYQKIWCVEPUPIL-QFIPXVFZSA-N
PubChem CID 127584
SMILES CCC1=C2CN3C(=CC4=C(C3=O)COC(=O)C4(CC)O)C2=NC5=CC=CC=C51

Specifications

Color Yellow
Quantity 100 mg
UN Number 1544
Formula Weight 376.41
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Ethylcamptothecin