missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Ethyl-3-indoleethanol 98.0+%, TCI America™
$208.78 - $829.51
Chemical Identifiers
| CAS | 41340-36-7 |
|---|---|
| Molecular Formula | C12H15NO |
| Molecular Weight (g/mol) | 189.26 |
| MDL Number | MFCD01718805 |
| InChI Key | UVSDNCAZVSQJQA-UHFFFAOYSA-N |
| Synonym | 7-Ethyl-3-(2-hydroxyethyl)indole, 7-Ethyltryptophol |
| PubChem CID | 198231 |
| IUPAC Name | 2-(7-ethyl-1H-indol-3-yl)ethan-1-ol |
| SMILES | CCC1=C2NC=C(CCO)C2=CC=C1 |
Chemical Identifiers
| 41340-36-7 | |
| 189.26 | |
| UVSDNCAZVSQJQA-UHFFFAOYSA-N | |
| 198231 | |
| CCC1=C2NC=C(CCO)C2=CC=C1 |
| C12H15NO | |
| MFCD01718805 | |
| 7-Ethyl-3-(2-hydroxyethyl)indole, 7-Ethyltryptophol | |
| 2-(7-ethyl-1H-indol-3-yl)ethan-1-ol |
Specifications
| 41340-36-7 | |
| White-Yellow | |
| MFCD01718805 | |
| 3077 | |
| UVSDNCAZVSQJQA-UHFFFAOYSA-N | |
| 2-(7-ethyl-1H-indol-3-yl)ethan-1-ol | |
| 198231 | |
| ≥98.0% (GC) | |
| 7-Ethyl-3-indoleethanol |
| 50°C | |
| C12H15NO | |
| 5 g | |
| 7-Ethyl-3-(2-hydroxyethyl)indole, 7-Ethyltryptophol | |
| CCC1=C2NC=C(CCO)C2=CC=C1 | |
| 189.26 | |
| 189.26 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : NL8512200
TSCA : No