missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Ethyl-2-methyl-4-undecanol 98.0+%, TCI America™
$129.10 - $1317.64
Chemical Identifiers
| CAS | 103-20-8 |
|---|---|
| Molecular Formula | C14H30O |
| Molecular Weight (g/mol) | 214.393 |
| MDL Number | MFCD00027253 |
| InChI Key | JKKBSZCOVNFRCQ-UHFFFAOYSA-N |
| Synonym | Tetradecanol, Tetradecyl Alcohol |
| PubChem CID | 97948 |
| IUPAC Name | 7-ethyl-2-methylundecan-4-ol |
| SMILES | CCCCC(CC)CCC(CC(C)C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E014425ML
|
TCI America
E014425ML |
25 mL |
Each for $129.10
|
|
|||||
|
E0144500ML
|
TCI America
E0144500ML |
500 mL |
Each for $1,317.64
|
|
|||||
Chemical Identifiers
| 103-20-8 | |
| 214.393 | |
| JKKBSZCOVNFRCQ-UHFFFAOYSA-N | |
| 97948 | |
| CCCCC(CC)CCC(CC(C)C)O |
| C14H30O | |
| MFCD00027253 | |
| Tetradecanol, Tetradecyl Alcohol | |
| 7-ethyl-2-methylundecan-4-ol |
Specifications
| 103-20-8 | |
| 264°C | |
| MFCD00027253 | |
| Tetradecanol, Tetradecyl Alcohol | |
| CCCCC(CC)CCC(CC(C)C)O | |
| 214.393 | |
| 214.39 | |
| Liquid |
| Colorless | |
| C14H30O | |
| 25 mL | |
| JKKBSZCOVNFRCQ-UHFFFAOYSA-N | |
| 7-ethyl-2-methylundecan-4-ol | |
| 97948 | |
| ≥98.0% (GC) | |
| 7-Ethyl-2-methyl-4-undecanol |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes