missing translation for 'onlineSavingsMsg'
Learn More

7-Ethyl-2-methyl-4-undecanol 98.0+%, TCI America™

Catalog No. E014425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E014425ML 25 mL
E0144500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E014425ML Supplier TCI America Supplier No. E014425ML

Chemical Identifiers

CAS 103-20-8
Molecular Formula C14H30O
Molecular Weight (g/mol) 214.393
MDL Number MFCD00027253
InChI Key JKKBSZCOVNFRCQ-UHFFFAOYSA-N
Synonym Tetradecanol, Tetradecyl Alcohol
PubChem CID 97948
IUPAC Name 7-ethyl-2-methylundecan-4-ol
SMILES CCCCC(CC)CCC(CC(C)C)O

Specifications

Color Colorless
Boiling Point 264°C
Quantity 25 mL
Formula Weight 214.39
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 7-Ethyl-2-methyl-4-undecanol
EINECSNumber (2)-0217
TSCA Yes