missing translation for 'onlineSavingsMsg'
Learn More

7-Ethyl-10-hydroxycamptothecin 98.0+%, TCI America™

Catalog No. p-7129129
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0748100MG 100 mg
E07481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0748100MG Supplier TCI America Supplier No. E0748100MG
Only null left

Chemical Identifiers

CAS 86639-52-3
Molecular Formula C22H20N2O5
Molecular Weight (g/mol) 392.411
MDL Number MFCD06762720
InChI Key FJHBVJOVLFPMQE-QFIPXVFZSA-N
Synonym 7-ethyl-10-hydroxycamptothecin, sn 38 lactone, 7-ethyl-10-hydroxy-camptothecin, 10-hydroxy-7-ethylcamptothecin, 7-ethyl-10-hydroxycamptothecine, le-sn38, 7-ethyl-10-hydroxy-20 s-camptothecin, captothecin, 7-ethyl-10-hydroxy, s-4,11-diethyl-4,9-dihydroxy-1h-pyrano 3',4':6,7 indolizino 1,2-b quinoline-3,14 4h,12h-dione, 7-ethyl-10-hydroxy camptothecin
PubChem CID 104842
ChEBI CHEBI:8988
SMILES CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)O

Specifications

Melting Point 217°C
Color Yellow
Quantity 100 mg
UN Number 1544
Formula Weight 392.41
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Ethyl-10-hydroxycamptothecin
RTECSNumber UQ0491000
TSCA No
Recommended Storage Freezer