Learn More
7-Ethyl-10-hydroxycamptothecin, 98%
CAS: 86639-52-3 | C22H20N2O5 | 392.4 g/mol
$144.95 - $144.95
Chemical Identifiers
| CAS | 86639-52-3 |
|---|---|
| Molecular Formula | C22H20N2O5 |
| Molecular Weight (g/mol) | 392.4 |
| InChI Key | FJHBVJOVLFPMQE-QFIPXVFZSA-N |
| Synonym | 7-ethyl-10-hydroxycamptothecin, sn 38 lactone, 7-ethyl-10-hydroxy-camptothecin, 10-hydroxy-7-ethylcamptothecin, 7-ethyl-10-hydroxycamptothecine, le-sn38, 7-ethyl-10-hydroxy-20 s-camptothecin, captothecin, 7-ethyl-10-hydroxy, s-4,11-diethyl-4,9-dihydroxy-1h-pyrano 3',4':6,7 indolizino 1,2-b quinoline-3,14 4h,12h-dione, 7-ethyl-10-hydroxy camptothecin |
| PubChem CID | 104842 |
| ChEBI | CHEBI:8988 |
| SMILES | CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC459091000
|
Thermo Scientific Chemicals
459091000 |
100 mg |
Each for $144.95
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 86639-52-3 | |
| 392.4 | |
| 7-ethyl-10-hydroxycamptothecin, sn 38 lactone, 7-ethyl-10-hydroxy-camptothecin, 10-hydroxy-7-ethylcamptothecin, 7-ethyl-10-hydroxycamptothecine, le-sn38, 7-ethyl-10-hydroxy-20 s-camptothecin, captothecin, 7-ethyl-10-hydroxy, s-4,11-diethyl-4,9-dihydroxy-1h-pyrano 3',4':6,7 indolizino 1,2-b quinoline-3,14 4h,12h-dione, 7-ethyl-10-hydroxy camptothecin | |
| CHEBI:8988 |
| C22H20N2O5 | |
| FJHBVJOVLFPMQE-QFIPXVFZSA-N | |
| 104842 | |
| CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)O |
Specifications
| 86639-52-3 | |
| 97.5% min. (HPLC) | |
| 100 mg | |
| FJHBVJOVLFPMQE-QFIPXVFZSA-N | |
| 392.4 | |
| CHEBI:8988 | |
| 98% |
| Authentic | |
| C22H20N2O5 | |
| 7-ethyl-10-hydroxycamptothecin, sn 38 lactone, 7-ethyl-10-hydroxy-camptothecin, 10-hydroxy-7-ethylcamptothecin, 7-ethyl-10-hydroxycamptothecine, le-sn38, 7-ethyl-10-hydroxy-20 s-camptothecin, captothecin, 7-ethyl-10-hydroxy, s-4,11-diethyl-4,9-dihydroxy-1h-pyrano 3',4':6,7 indolizino 1,2-b quinoline-3,14 4h,12h-dione, 7-ethyl-10-hydroxy camptothecin | |
| CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)O | |
| 104842 | |
| 392.4 | |
| 7-Ethyl-10-hydroxycamptothecin |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
Do not eat,drink or smoke when us
GHS Signal Word: Danger
RUO – Research Use Only