missing translation for 'onlineSavingsMsg'
Learn More

7-(Diethylamino)coumarin-3-carbonitrile 98.0+%, TCI America™

Catalog No. D44625G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44625G 5 g
D44621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44625G Supplier TCI America Supplier No. D44625G
Only null left

Chemical Identifiers

CAS 51473-74-6
Molecular Formula C14H14N2O2
Molecular Weight (g/mol) 242.278
MDL Number MFCD00068688
InChI Key LOUYEVRVQGFIFB-UHFFFAOYSA-N
Synonym 3-Cyano-7-(diethylamino)coumarin
PubChem CID 688937
IUPAC Name 7-(diethylamino)-2-oxochromene-3-carbonitrile
SMILES CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C#N

Specifications

Melting Point 230°C
Color Yellow
Quantity 5 g
Formula Weight 242.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Diethylamino)coumarin-3-carbonitrile