missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(Diethylamino)coumarin-3-carbonitrile 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 230°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 242.28 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 7-(Diethylamino)coumarin-3-carbonitrile |
Safety and Handling
| TSCA | No |