missing translation for 'onlineSavingsMsg'
Learn More

7-(Diethylamino)-4-(trifluoromethyl)coumarin 98.0+%, TCI America™

Catalog No. D4466200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4466200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4466200MG Supplier TCI America Supplier No. D4466200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41934-47-8
Molecular Formula C14H14F3NO2
Molecular Weight (g/mol) 285.266
MDL Number MFCD00057313
InChI Key UIMOXRDVWDLOHW-UHFFFAOYSA-N
Synonym Coumarin 481
PubChem CID 94523
ChEBI CHEBI:51777
IUPAC Name 7-(diethylamino)-4-(trifluoromethyl)chromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C(=CC(=O)O2)C(F)(F)F

Specifications

Quantity 200 mg
Formula Weight 285.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Diethylamino)-4-(trifluoromethyl)coumarin