missing translation for 'onlineSavingsMsg'
Learn More

7-Deazahypoxanthine 98.0+%, TCI America™

Supplier:  TCI America D43241G

Catalog No. D43241G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7-Deazahypoxanthine
345°C
MFCD08445809
7h-pyrrolo 2,3-d pyrimidin-4-ol, pyrrolo 2,3-d pyrimidin-4-ol, 7-deazahypoxanthine, 3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 4-hydroxypyrrolo 2,3-d pyrimidine, 4-hydroxypyrrolopyrimidine, 1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 3,7-dihydro-4h-pyrrolo 2,3-d pyrimidin-4-one, pyrrolo 2,3-d pyrimidin-4-one
O=C1N=CNC2=C1C=CN2
135.13
135.13
Crystalline Powder
3680-71-5
C6H5N3O
1 g
FBMZEITWVNHWJW-UHFFFAOYSA-N
1H,4H,7H-pyrrolo[2,3-d]pyrimidin-4-one
96194
≥98.0% (HPLC,N)

Chemical Identifiers

3680-71-5
135.13
FBMZEITWVNHWJW-UHFFFAOYSA-N
96194
O=C1N=CNC2=C1C=CN2
C6H5N3O
MFCD08445809
7h-pyrrolo 2,3-d pyrimidin-4-ol, pyrrolo 2,3-d pyrimidin-4-ol, 7-deazahypoxanthine, 3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 4-hydroxypyrrolo 2,3-d pyrimidine, 4-hydroxypyrrolopyrimidine, 1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 3,7-dihydro-4h-pyrrolo 2,3-d pyrimidin-4-one, pyrrolo 2,3-d pyrimidin-4-one
1H,4H,7H-pyrrolo[2,3-d]pyrimidin-4-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UY9440000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions