missing translation for 'onlineSavingsMsg'
Learn More

7-Deazahypoxanthine, 97%

Catalog No. AAH6453706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6453706 5 g
AAH6453714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6453706 Supplier Thermo Scientific Chemicals Supplier No. H6453706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3680-71-5 | C6H5N3O | 135.13 g/mol

It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Moisture and light sensitive. Store at -20°C

Chemical Identifiers

CAS 3680-71-5
Molecular Formula C6H5N3O
Molecular Weight (g/mol) 135.13
MDL Number MFCD08445809
InChI Key FBMZEITWVNHWJW-UHFFFAOYSA-N
Synonym 7h-pyrrolo 2,3-d pyrimidin-4-ol, pyrrolo 2,3-d pyrimidin-4-ol, 7-deazahypoxanthine, 3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 4-hydroxypyrrolo 2,3-d pyrimidine, 4-hydroxypyrrolopyrimidine, 1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 3,7-dihydro-4h-pyrrolo 2,3-d pyrimidin-4-one, pyrrolo 2,3-d pyrimidin-4-one
PubChem CID 96194
IUPAC Name 1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
SMILES O=C1N=CNC2=C1C=CN2

Specifications

Melting Point 345°C to 350°C
Color Brown
Quantity 5 g
Beilstein 607869
Sensitivity Light and moisture sensitive
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 135.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 7-Deazahypoxanthine
Hazard Category H319-H334
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P280i-P285-P304+P341-P305+P351+P338-P342+P311-P501c
RTECSNumber UY9440000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only