missing translation for 'onlineSavingsMsg'
Learn More

7-Deazaguanine, 97%, Thermo Scientific™

Catalog No. p-7036323
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54595MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH54595MD Supplier Thermo Scientific Chemicals Supplier No. H54595MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethidium bromide efficiently intercalates into synthetic duplex DNAs with 7-deazaguanine in place of guanine, but shows none of the fluorescence enhancement usually observed upon intercalation. 7-Deazaadenine-duplexes show about 70% of the expected fluorescence enhancement.

Solubility
Soluble in N,N-Dimethylformamide, Dimethyl sulfoxide.

Notes
Store at room temperature. Keep away from oxidizing agents and acids.

Chemical Identifiers

CAS 7355-55-7
Molecular Formula C6H6N4O
Molecular Weight (g/mol) 150.14
MDL Number MFCD09033259
InChI Key OLAFFPNXVJANFR-UHFFFAOYSA-N
Synonym 7-deazaguanine, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, unii-gpl8t5zo3m, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, gpl8t5zo3m, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one
PubChem CID 96253
IUPAC Name 2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
SMILES NC1=NC(=O)C2=C(NC=C2)N1

Specifications

Quantity 250 mg
Beilstein 512680
Solubility Information Soluble in N,N-Dimethylformamide,Dimethyl sulfoxide.
Formula Weight 150.14
Percent Purity 97%
Chemical Name or Material 7-Deazaguanine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures