missing translation for 'onlineSavingsMsg'
Learn More

7-Chlorokynurenic acid, 98+%

NMDA receptor antagonist at the glycine site | CAS: 18000-24-3 | C10H6ClNO3 | 223.61 g/mol

$353.05 - $660.22

Chemical Identifiers

CAS 18000-24-3
Molecular Formula C10H6ClNO3
Molecular Weight (g/mol) 223.61
MDL Number MFCD00069227
InChI Key UAWVRVFHMOSAPU-UHFFFAOYSA-N
Synonym 7-chlorokynurenic acid, 7-chloro-4-hydroxyquinoline-2-carboxylic acid, 7-cl-kyna, 7-chloro-kynurenic acid, unii-s7936qon2k, 7-chloro-4-hydroxy-2-carboxyquinoline, 7-cka, 7-chloro-4-hydroxy-2-quinolinecarboxylic acid, 2-quinolinecarboxylic acid, 7-chloro-1,4-dihydro-4-oxo, cka
PubChem CID 1884
ChEBI CHEBI:63965
IUPAC Name 7-chloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)C2=CC=C(Cl)C=C2N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61359MA
View Documents
Thermo Scientific Chemicals
J61359MA
10 mg
Each for $353.05
Only null left
 
AAJ61359MF
View Documents
Thermo Scientific Chemicals
J61359MF
50 mg
Each for $660.22
Only null left
 
Description

Description

NMDA receptor antagonist at the glycine site

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18000-24-3
223.61
UAWVRVFHMOSAPU-UHFFFAOYSA-N
1884
7-chloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
C10H6ClNO3
MFCD00069227
7-chlorokynurenic acid, 7-chloro-4-hydroxyquinoline-2-carboxylic acid, 7-cl-kyna, 7-chloro-kynurenic acid, unii-s7936qon2k, 7-chloro-4-hydroxy-2-carboxyquinoline, 7-cka, 7-chloro-4-hydroxy-2-quinolinecarboxylic acid, 2-quinolinecarboxylic acid, 7-chloro-1,4-dihydro-4-oxo, cka
CHEBI:63965
OC(=O)C1=CC(=O)C2=CC=C(Cl)C=C2N1

Specifications

18000-24-3
C10H6ClNO3
10 mg
Soluble in DMSO or dilute sodium hydroxide
OC(=O)C1=CC(=O)C2=CC=C(Cl)C=C2N1
223.61
CHEBI:63965
≥99%
7-Chlorokynurenic acid
White to Yellow
MFCD00069227
7-chlorokynurenic acid, 7-chloro-4-hydroxyquinoline-2-carboxylic acid, 7-cl-kyna, 7-chloro-kynurenic acid, unii-s7936qon2k, 7-chloro-4-hydroxy-2-carboxyquinoline, 7-cka, 7-chloro-4-hydroxy-2-quinolinecarboxylic acid, 2-quinolinecarboxylic acid, 7-chloro-1,4-dihydro-4-oxo, cka
UAWVRVFHMOSAPU-UHFFFAOYSA-N
7-chloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
1884
223.62
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 241-913-6

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only