missing translation for 'onlineSavingsMsg'
Learn More

7-Chlorokynurenic acid, 98+%

Catalog No. AAJ61359MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61359MF 50 mg
AAJ61359MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61359MF Supplier Thermo Scientific Chemicals Supplier No. J61359MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

NMDA receptor antagonist at the glycine site | CAS: 18000-24-3 | C10H6ClNO3 | 223.61 g/mol

NMDA receptor antagonist at the glycine site

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18000-24-3
Molecular Formula C10H6ClNO3
Molecular Weight (g/mol) 223.61
MDL Number MFCD00069227
InChI Key UAWVRVFHMOSAPU-UHFFFAOYSA-N
Synonym 7-chlorokynurenic acid, 7-chloro-4-hydroxyquinoline-2-carboxylic acid, 7-cl-kyna, 7-chloro-kynurenic acid, unii-s7936qon2k, 7-chloro-4-hydroxy-2-carboxyquinoline, 7-cka, 7-chloro-4-hydroxy-2-quinolinecarboxylic acid, 2-quinolinecarboxylic acid, 7-chloro-1,4-dihydro-4-oxo, cka
PubChem CID 1884
ChEBI CHEBI:63965
IUPAC Name 7-chloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)C2=CC=C(Cl)C=C2N1

Specifications

Color White to Yellow
Quantity 50 mg
Solubility Information Soluble in DMSO or dilute sodium hydroxide
Formula Weight 223.62
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 7-Chlorokynurenic acid
EINECSNumber 241-913-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only