missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-6-nitro-4-hydroxyquinazoline 98.0+%, TCI America™

Catalog No. C23325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23325G 5 g
C233225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23325G Supplier TCI America Supplier No. C23325G
Only null left

Chemical Identifiers

CAS 53449-14-2
Molecular Formula C8H4ClN3O3
Molecular Weight (g/mol) 225.588
MDL Number MFCD07772904
InChI Key URDYTQYZXZKBQT-UHFFFAOYSA-N
Synonym 7-chloro-6-nitroquinazolin-4 3h-one, 7-chloro-4-hydroxy-6-nitroquinazoline, 7-chloro-6-nitro-4-hydroxyquinazoline, 6-nitro-7-chloro-4-hydroxyquinazoline, 7-chloro-6-nitroquinazolin-4-ol, 7-chloro-6-nitroquinazolin-4-one, 7-chloro-6-nitro-quinazolin-4-one, 7-chloro-6-nitro-3h-quinazolin-4-one, 4 1h-quinazolinone, 7-chloro-6-nitro, 7-chloro-6-nitro-3,4-dihydroquinazolin-4-one
PubChem CID 12441237
IUPAC Name 7-chloro-6-nitro-1H-quinazolin-4-one
SMILES C1=C2C(=CC(=C1[N+](=O)[O-])Cl)NC=NC2=O

Specifications

Melting Point 303°C
Quantity 5 g
Formula Weight 225.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Chloro-6-nitro-4-hydroxyquinazoline