missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-6-nitro-4(3H)-quinazolinone, 97%, Thermo Scientific™

Chemical Identifiers

CAS 53449-14-2
Molecular Formula C8H4ClN3O3
Molecular Weight (g/mol) 225.59
InChI Key URDYTQYZXZKBQT-UHFFFAOYSA-N
Synonym 7-chloro-6-nitroquinazolin-4 3h-one, 7-chloro-4-hydroxy-6-nitroquinazoline, 7-chloro-6-nitro-4-hydroxyquinazoline, 6-nitro-7-chloro-4-hydroxyquinazoline, 7-chloro-6-nitroquinazolin-4-ol, 7-chloro-6-nitroquinazolin-4-one, 7-chloro-6-nitro-quinazolin-4-one, 7-chloro-6-nitro-3h-quinazolin-4-one, 4 1h-quinazolinone, 7-chloro-6-nitro, 7-chloro-6-nitro-3,4-dihydroquinazolin-4-one
PubChem CID 12441237
IUPAC Name 7-chloro-6-nitro-1H-quinazolin-4-one
SMILES C1=C2C(=CC(=C1[N+](=O)[O-])Cl)NC=NC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC450790050
View Documents
Thermo Scientific Chemicals
450790050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC450790250
SDS View Documents
Thermo Scientific Chemicals
450790250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

53449-14-2
225.59
7-chloro-6-nitroquinazolin-4 3h-one, 7-chloro-4-hydroxy-6-nitroquinazoline, 7-chloro-6-nitro-4-hydroxyquinazoline, 6-nitro-7-chloro-4-hydroxyquinazoline, 7-chloro-6-nitroquinazolin-4-ol, 7-chloro-6-nitroquinazolin-4-one, 7-chloro-6-nitro-quinazolin-4-one, 7-chloro-6-nitro-3h-quinazolin-4-one, 4 1h-quinazolinone, 7-chloro-6-nitro, 7-chloro-6-nitro-3,4-dihydroquinazolin-4-one
7-chloro-6-nitro-1H-quinazolin-4-one
C8H4ClN3O3
URDYTQYZXZKBQT-UHFFFAOYSA-N
12441237
C1=C2C(=CC(=C1[N+](=O)[O-])Cl)NC=NC2=O

Specifications

53449-14-2
Glass bottle
5g
Solubility in water: .. Other solubilities: slightly soluble in dmf
C1=C2C(=CC(=C1[N+](=O)[O-])Cl)NC=NC2=O
225.59
225.59
7-Chloro-6-nitro-4(3H)-quinazolinone
96% min. (HPLC)
C8H4ClN3O3
7-chloro-6-nitroquinazolin-4 3h-one, 7-chloro-4-hydroxy-6-nitroquinazoline, 7-chloro-6-nitro-4-hydroxyquinazoline, 6-nitro-7-chloro-4-hydroxyquinazoline, 7-chloro-6-nitroquinazolin-4-ol, 7-chloro-6-nitroquinazolin-4-one, 7-chloro-6-nitro-quinazolin-4-one, 7-chloro-6-nitro-3h-quinazolin-4-one, 4 1h-quinazolinone, 7-chloro-6-nitro, 7-chloro-6-nitro-3,4-dihydroquinazolin-4-one
URDYTQYZXZKBQT-UHFFFAOYSA-N
7-chloro-6-nitro-1H-quinazolin-4-one
12441237
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only