missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid 98.0+%, TCI America™
$65.24 - $171.49
Chemical Identifiers
| CAS | 86393-33-1 |
|---|---|
| Molecular Formula | C13H8ClFNO3 |
| Molecular Weight (g/mol) | 280.66 |
| MDL Number | MFCD00792458 |
| InChI Key | ISPVACVJFUIDPD-UHFFFAOYSA-M |
| Synonym | Fluoroquinolonic Acid |
| PubChem CID | 483180 |
| IUPAC Name | 7-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate |
| SMILES | [O-]C(=O)C1=CN(C2CC2)C2=CC(Cl)=C(F)C=C2C1=O |
Chemical Identifiers
| 86393-33-1 | |
| 280.66 | |
| ISPVACVJFUIDPD-UHFFFAOYSA-M | |
| 483180 | |
| [O-]C(=O)C1=CN(C2CC2)C2=CC(Cl)=C(F)C=C2C1=O |
| C13H8ClFNO3 | |
| MFCD00792458 | |
| Fluoroquinolonic Acid | |
| 7-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate |
Specifications
| 86393-33-1 | |
| Yellow | |
| MFCD00792458 | |
| Fluoroquinolonic Acid | |
| [O-]C(=O)C1=CN(C2CC2)C2=CC(Cl)=C(F)C=C2C1=O | |
| 280.66 | |
| 281.67 | |
| Crystalline Powder |
| 244°C | |
| C13H8ClFNO3 | |
| 5 g | |
| ISPVACVJFUIDPD-UHFFFAOYSA-M | |
| 7-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate | |
| 483180 | |
| ≥98.0% (HPLC,T) | |
| 7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid |
Safety and Handling
RTECSNumber : VB1993795
TSCA : No