missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoquinoline 98.0+%, TCI America™

Catalog No. B41551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41551G 1 g
B41555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41551G Supplier TCI America Supplier No. B41551G

Chemical Identifiers

CAS 4965-36-0
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD03695823
InChI Key XYBSZCUHOLWQQU-UHFFFAOYSA-N
Synonym quinoline, 7-bromo, 7-bromo-quinoline, pubchem12788, ksc238o9n, abbypharma ap-12-10883
PubChem CID 521259
IUPAC Name 7-bromoquinoline
SMILES C1=CC2=C(C=C(C=C2)Br)N=C1

Specifications

Melting Point 35°C
Color White-Yellow
Quantity 1 g
Formula Weight 208.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 7-Bromoquinoline
TSCA No
Recommended Storage Refrigerator