missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoquinoline, 95%

Catalog No. p-7055412
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6398703 1 g
AAH6398706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6398703 Supplier Thermo Scientific Chemicals Supplier No. H6398703
Only null left

CAS: 4965-36-0 | C9H6BrN | 208.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4965-36-0
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD03695823
InChI Key XYBSZCUHOLWQQU-UHFFFAOYSA-N
Synonym quinoline, 7-bromo, 7-bromo-quinoline, pubchem12788, ksc238o9n, abbypharma ap-12-10883
PubChem CID 521259
IUPAC Name 7-bromoquinoline
SMILES C1=CC2=C(C=C(C=C2)Br)N=C1

Specifications

Quantity 1 g
Formula Weight 208.06
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 7-Bromoquinoline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only