missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoisatin 97.0+%, TCI America™

Catalog No. B39741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39741G 1 g
B39745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39741G Supplier TCI America Supplier No. B39741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20780-74-9
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00774354
InChI Key OCVKSIWBTJCXPV-UHFFFAOYSA-N
Synonym 7-bromoisatin, 7-bromoindoline-2,3-dione, 7-bromo-2,3-dioxoindoline, 1h-indole-2,3-dione, 7-bromo, 7-bromoindole-2,3-dione, 7-bromo-2,3-dihydro-1h-indole-2,3-dione, 7-bromoindole-1h-2,3-dione, 7-bromo-2,3-indolinedione, 7-bromo isatin, 7-bromo-isatin
PubChem CID 2302353
IUPAC Name 7-bromo-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)Br)NC(=O)C2=O

Specifications

Melting Point 200°C
Quantity 1 g
Formula Weight 226.03
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromoisatin