missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoindole 97.0+%, TCI America™

Catalog No. B28735G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28735G 5 g
B28731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28735G missing translation for 'mfr' TCI America missing translation for 'supplierNo' B28735G

Chemical Identifiers

CAS 51417-51-7
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00799492
InChI Key RDSVSEFWZUWZHW-UHFFFAOYSA-N
Synonym 7-bromoindole, 1h-indole, 7-bromo, 7-bromo-indole, 7-bromindol, 7-bromoindol, 7-brom-1h-indole, pubchem7318, acmc-209ktm, ksc269i1f
PubChem CID 2757020
IUPAC Name 7-bromo-1H-indole
SMILES C1=CC2=C(C(=C1)Br)NC=C2

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 5 g
Formula Weight 196.05
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromoindole