missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-5-fluorobenzo[b]furan, 97%

Catalog No. AAH33948MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33948MD 250 mg
AAH3394803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33948MD Supplier Thermo Scientific Chemicals Supplier No. H33948MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 253429-19-5 | C8H4BrFO | 215.021 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 253429-19-5
Molecular Formula C8H4BrFO
Molecular Weight (g/mol) 215.021
MDL Number MFCD09056780
InChI Key TYBMMMOEVBNTDU-UHFFFAOYSA-N
PubChem CID 22144816
IUPAC Name 7-bromo-5-fluoro-1-benzofuran
SMILES C1=COC2=C(C=C(C=C21)F)Br

Specifications

Quantity 250 mg
Synonym 7-bromo-5-fluorobenzofuran, 7-bromo-5-fluorobenzo b furan, 5-fluoro-7-bromobenzofuran, 7-bromo-5-fluoro-benzofuran, benzofuran,7-bromo-5-fluoro, benzofuran, 7-bromo-5-fluoro, 7-bromanyl-5-fluoranyl-1-benzofuran
Formula Weight 215.02
Percent Purity 97%
Chemical Name or Material 7-Bromo-5-fluorobenzo[b]furan
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only