missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals

Catalog No. p-7041078
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6102103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6102103 Supplier Thermo Scientific Chemicals Supplier No. H6102103
Only null left

7-Bromo-3,4-dihydro-2H-1,4-benzothiazine used in many areas of medicine. They are used in industries as dyes and as antioxidants of rubber and natural elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine used in many areas of medicine. They are used in industries as dyes and as antioxidants of rubber and natural elastomers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 193414-60-7
Molecular Formula C8H8BrNS
Molecular Weight (g/mol) 230.12
MDL Number MFCD11847754
InChI Key QWWYGAWGUCQKDL-UHFFFAOYSA-N
Synonym 7-bromo-3,4-dihydro-2h-benzo b 1,4 thiazine
PubChem CID 83823811
IUPAC Name 7-bromo-3,4-dihydro-2H-1,4-benzothiazine
SMILES BrC1=CC2=C(NCCS2)C=C1

Specifications

Quantity 1 g
Formula Weight 230.12
Percent Purity 97%
Chemical Name or Material 7-Bromo-3,4-dihydro-2H-1,4-benzothiazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only