missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-3,4-dihydro-1,5-benzodioxepin, 96%

Catalog No. AAH2759403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2759403 1 g
AAH2759406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2759403 Supplier Thermo Scientific Chemicals Supplier No. H2759403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 147644-11-9 | C9H9BrO2 | 229.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 147644-11-9
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00508291
InChI Key AZCHNKNSOZJHSH-UHFFFAOYSA-N
PubChem CID 2776394
IUPAC Name 7-bromo-3,4-dihydro-2H-1,5-benzodioxepine
SMILES C1COC2=C(C=C(C=C2)Br)OC1

Specifications

Boiling Point 145°C to 148°C (16 mmHg)
Flash Point >120°C (248°F)
Refractive Index 1.579
Quantity 1 g
Synonym 7-bromo-3,4-dihydro-2h-benzo b 1,4 dioxepine, 2h-1,5-benzodioxepin, 7-bromo-3,4-dihydro, acmc-1c5pl, 7-bromo-2h,3h,4h-benzo b 1,4-dioxepin, 7-bromo-3,4-dihydro-2h-1,5-benzodioxepin, 2h-1,5-benzodioxepin,7-bromo-3,4-dihydro, 7-bromo-3,4-dihydro-1,5-benzodioxepin
Formula Weight 229.07
Percent Purity 96%
Chemical Name or Material 7-Bromo-3,4-dihydro-1,5-benzodioxepin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only