missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde 96.0+%, TCI America™
$169.63 - $596.49
Chemical Identifiers
| CAS | 852054-42-3 |
|---|---|
| Molecular Formula | C7H5BrO3S |
| Molecular Weight (g/mol) | 249.078 |
| MDL Number | MFCD08690306 |
| InChI Key | YEKBJVHBVICUOZ-UHFFFAOYSA-N |
| Synonym | 7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbaldehyde, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxine-5-carbaldehyde, 7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxaldehyde, 2,3-dihydro-7-bromothieno 3,4-b 1,4 dioxin-5-carbaldehyde, 7-bromo-2,3-dihydrothieno 3,4-b-1,4-dioxin-5-carbaldehyde, 7-bromo-2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbaldehyde, thieno 3,4-b-1,4-dioxin-5-carboxaldehyde,7-bromo-2,3-dihydro |
| PubChem CID | 11391008 |
| IUPAC Name | 5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-7-carbaldehyde |
| SMILES | C1COC2=C(SC(=C2O1)C=O)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5396200MG
|
TCI America
B5396200MG |
200 mg |
Each for $169.63
|
|
|||||
|
B53961G
|
TCI America
B53961G |
1 g |
Each for $596.49
|
|
|||||
Chemical Identifiers
| 852054-42-3 | |
| 249.078 | |
| YEKBJVHBVICUOZ-UHFFFAOYSA-N | |
| 11391008 | |
| C1COC2=C(SC(=C2O1)C=O)Br |
| C7H5BrO3S | |
| MFCD08690306 | |
| 7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbaldehyde, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxine-5-carbaldehyde, 7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxaldehyde, 2,3-dihydro-7-bromothieno 3,4-b 1,4 dioxin-5-carbaldehyde, 7-bromo-2,3-dihydrothieno 3,4-b-1,4-dioxin-5-carbaldehyde, 7-bromo-2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbaldehyde, thieno 3,4-b-1,4-dioxin-5-carboxaldehyde,7-bromo-2,3-dihydro | |
| 5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-7-carbaldehyde |
Specifications
| 852054-42-3 | |
| Gray-White | |
| MFCD08690306 | |
| 7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbaldehyde, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxine-5-carbaldehyde, 7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxaldehyde, 2,3-dihydro-7-bromothieno 3,4-b 1,4 dioxin-5-carbaldehyde, 7-bromo-2,3-dihydrothieno 3,4-b-1,4-dioxin-5-carbaldehyde, 7-bromo-2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbaldehyde, thieno 3,4-b-1,4-dioxin-5-carboxaldehyde,7-bromo-2,3-dihydro | |
| C1COC2=C(SC(=C2O1)C=O)Br | |
| 249.078 | |
| 249.08 | |
| Crystalline Powder |
| 149°C | |
| C7H5BrO3S | |
| 200 mg | |
| YEKBJVHBVICUOZ-UHFFFAOYSA-N | |
| 5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-7-carbaldehyde | |
| 11391008 | |
| ≥96.0% (GC) | |
| 7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde |
Safety and Handling
TSCA : No