missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-methyl-3,4-dihydro-2(1H)-quinolinone, 96%

Catalog No. AAH33197MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33197MD 250 mg
AAH3319703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33197MD Supplier Thermo Scientific Chemicals Supplier No. H33197MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1086386-20-0 | C10H10BrNO | 240.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1086386-20-0
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.10
MDL Number MFCD11655616
InChI Key XVBPEXCFWANWIM-UHFFFAOYSA-N
Synonym 7-bromo-1-methyl-3,4-dihydro-2 1h-quinolinone, 7-bromo-1-methyl-3,4-dihydroquinolin-2 1h-one, 2 1h-quinolinone, 7-bromo-3,4-dihydro-1-methyl
PubChem CID 46741415
IUPAC Name 7-bromo-1-methyl-3,4-dihydroquinolin-2-one
SMILES CN1C(=O)CCC2=C1C=C(Br)C=C2

Specifications

Melting Point 130°C to 134°C
Quantity 250 mg
Formula Weight 240.1
Percent Purity 96%
Chemical Name or Material 7-Bromo-1-methyl-3,4-dihydro-2(1H)-quinolinone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only