missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-heptanol 88.0+%, TCI America™

Catalog No. B18521G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18521G 1 g
B18525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18521G Supplier TCI America Supplier No. B18521G

Chemical Identifiers

CAS 10160-24-4
Molecular Formula C7H15BrO
Molecular Weight (g/mol) 195.1
MDL Number MFCD00041708
InChI Key MMXRRNUXCHUHOE-UHFFFAOYSA-N
Synonym 7-bromo-1-heptanol, 1-heptanol, 7-bromo, 7-bromoheptanol, 7-bromoheptane-1-ol
PubChem CID 66284
IUPAC Name 7-bromoheptan-1-ol
SMILES C(CCCO)CCCBr

Specifications

Quantity 1 g
Formula Weight 195.10
Percent Purity ≥88.0% (GC)
Physical Form Liquid
Chemical Name or Material 7-Bromo-1-heptanol
TSCA Yes
Recommended Storage Freezer