missing translation for 'onlineSavingsMsg'
Learn More

7-Benzyloxyindole 98.0+%, TCI America™

Catalog No. B28465G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28465G Supplier TCI America Supplier No. B28465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20289-27-4
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.28
MDL Number MFCD00037974
InChI Key DIGZMTAFOACVBW-UHFFFAOYSA-N
Synonym 7-benzyloxyindole, 7-benzyloxy-1h-indole, 7-phenylmethoxy indole, 1h-indole, 7-phenylmethoxy, 7-phenylmethoxy-1h-indole, 7-benzyloxy-1h-indol, 7-benzyloxindole, 7-benzyloxy indole
PubChem CID 260798
IUPAC Name 7-(benzyloxy)-1H-indole
SMILES C(OC1=C2NC=CC2=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 73°C
Quantity 5 g
Formula Weight 223.28
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 7-Benzyloxyindole