missing translation for 'onlineSavingsMsg'
Learn More

7-Benzyloxyindole, 98%

Catalog No. AAH2577614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2577614 25 g
AAH2577603 1 g
AAH2577606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2577614 Supplier Thermo Scientific Chemicals Supplier No. H2577614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20289-27-4 | C15H13NO | 223.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20289-27-4
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.28
MDL Number MFCD00037974
InChI Key DIGZMTAFOACVBW-UHFFFAOYSA-N
Synonym 7-benzyloxyindole, 7-benzyloxy-1h-indole, 7-phenylmethoxy indole, 1h-indole, 7-phenylmethoxy, 7-phenylmethoxy-1h-indole, 7-benzyloxy-1h-indol, 7-benzyloxindole, 7-benzyloxy indole
PubChem CID 260798
IUPAC Name 7-phenylmethoxy-1H-indole
SMILES C(OC1=C2NC=CC2=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 70°C to 74°C
Quantity 25 g
Formula Weight 223.28
Percent Purity 98%
Chemical Name or Material 7-Benzyloxyindole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only