missing translation for 'onlineSavingsMsg'
Learn More

7-Benzyloxy-6-methoxy-4(3H)-quinazolinone, 96%

Catalog No. AAH3413106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3413106 5 g
AAH3413103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3413106 Supplier Thermo Scientific Chemicals Supplier No. H3413106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 179688-01-8 | C16H14N2O3 | 282.299 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 179688-01-8
Molecular Formula C16H14N2O3
Molecular Weight (g/mol) 282.299
MDL Number MFCD04115119
InChI Key ZCUFFSHMOAEEIL-UHFFFAOYSA-N
Synonym 7-benzyloxy-6-methoxyquinazolin-4 3h-one, 6-methoxy-7-benzyloxyquinazolin-4-one, 7-benzyloxy-6-methoxy-3h-quinazolin-4-one, 7-benzyloxy-6-methoxyquinazolin-4-ol, 7-benzyloxy-6-methoxyquinazolin-4 1h-one, 4 3h-quinazolinone, 6-methoxy-7-phenylmethoxy, 7-benzyloxy-6-methoxy-4 3h-quinazolinone, 4 1h-quinazolinone, 6-methoxy-7-phenylmethoxy
PubChem CID 10540849
IUPAC Name 6-methoxy-7-phenylmethoxy-1H-quinazolin-4-one
SMILES COC1=C(C=C2C(=C1)C(=O)N=CN2)OCC3=CC=CC=C3

Specifications

Quantity 5 g
Formula Weight 284.31
Percent Purity 96%
Chemical Name or Material 7-Benzyloxy-6-methoxy-4(3H)-quinazolinone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only