missing translation for 'onlineSavingsMsg'
Learn More

7-Benzyloxy-6-methoxy-3H-quinazolin-4-one 98.0+%, TCI America™

Catalog No. B43175G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43175G 5 g
B43171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43175G Supplier TCI America Supplier No. B43175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 179688-01-8
Molecular Formula C16H14N2O3
Molecular Weight (g/mol) 282.299
MDL Number MFCD04115119
InChI Key ZCUFFSHMOAEEIL-UHFFFAOYSA-N
Synonym 7-benzyloxy-6-methoxyquinazolin-4 3h-one, 6-methoxy-7-benzyloxyquinazolin-4-one, 7-benzyloxy-6-methoxy-3h-quinazolin-4-one, 7-benzyloxy-6-methoxyquinazolin-4-ol, 7-benzyloxy-6-methoxyquinazolin-4 1h-one, 4 3h-quinazolinone, 6-methoxy-7-phenylmethoxy, 7-benzyloxy-6-methoxy-4 3h-quinazolinone, 4 1h-quinazolinone, 6-methoxy-7-phenylmethoxy
PubChem CID 10540849
IUPAC Name 6-methoxy-7-phenylmethoxy-1H-quinazolin-4-one
SMILES COC1=C(C=C2C(=C1)C(=O)N=CN2)OCC3=CC=CC=C3

Specifications

Melting Point 269°C
Color White-Yellow
Quantity 5 g
Formula Weight 282.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Benzyloxy-6-methoxy-3H-quinazolin-4-one